2-Cyano-6-fluorobenzoic acid ≥95%

Supplier: Matrix Scientific

068865-250MG
101938-002EA 538.46 USD
101938-002
2-Cyano-6-fluorobenzoic acid ≥95%
2-Cyano-6-fluorobenzoic acid

Hazard: Irritant

Formula: C₈H₄FNO₂
MDL Number: MFCD07368776
CAS Number: 887266-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR