2,2,2-Trifluoro-N-(6-iodoimidazo[1,2-a]pyridin-2-yl)acetamide ≥95%

Supplier: Matrix Scientific

Synonyms: 2,2,2-trifluoro-N-{6-iodoimidazo[1,2-a]pyridin-2-yl}acetamide

068856-1G
101937-960EA 989.23 USD
101937-960
2,2,2-Trifluoro-N-(6-iodoimidazo[1,2-a]pyridin-2-yl)acetamide ≥95%
2,2,2-Trifluoro-N-(6-iodoimidazo[1,2-a]pyridin-2-yl)acetamide

Hazard: Irritant

Formula: C₉H₅F₃IN₃O
MW: 355.06 g/mol
MDL Number: MFCD11878598
CAS Number: 209971-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR