(±)-Cbz-α-phosphonoglycine trimethyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-{[(benzyloxy)carbonyl]amino}-2-(dimethoxyphosphoryl)acetateMethyl 2-(((benzyloxy)carbonyl)amino)-2-(dimethoxyphosphoryl)acetate(±)-Methyl 2-benzyloxycarbonylamino-2-(dimethoxyphosphinyl)acetate(±)-Benzyloxycarbonyl-α-phosphonoglycine trimethyl ester

068855-5G
101937-956EA 26.45 USD
101937-956
(±)-Cbz-α-phosphonoglycine trimethyl ester ≥95%
(±)-Cbz-α-phosphonoglycine trimethyl ester

Hazard: Irritant

Formula: C₁₃H₁₈NO₇P
MW: 331.27 g/mol
MDL Number: MFCD00043304
CAS Number: 88568-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR