4-Methyl-1H-pyrrolo[2,3-b]pyridine ≥95%

Supplier: Matrix Scientific

068852-250MG
101937-942EA 669.23 USD
101937-942
4-Methyl-1H-pyrrolo[2,3-b]pyridine ≥95%
4-Methyl-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₈H₈N₂
MDL Number: MFCD00956196
CAS Number: 824-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR