cis-3-(Aminomethyl)cyclobutanol ≥95%

Supplier: Matrix Scientific

068850-500MG
101937-934EA 1347.69 USD
101937-934
cis-3-(Aminomethyl)cyclobutanol ≥95%
cis-3-(Aminomethyl)cyclobutanol

Hazard: Irritant

Formula: C₅H₁₁NO
MDL Number: MFCD16661176
CAS Number: 917827-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR