Pyrrolo[1,2-b]pyridazin-4(1H)-one ≥95%

Supplier: Matrix Scientific

068847-500MG
101937-922EA 1183.08 USD
101937-922
Pyrrolo[1,2-b]pyridazin-4(1H)-one ≥95%
Pyrrolo[1,2-b]pyridazin-4(1H)-one

Hazard: Irritant

Formula: C₇H₆N₂O
MDL Number: MFCD12923493
CAS Number: 888720-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR