2-Amino-4-methylthiophene ≥95%

Supplier: Matrix Scientific

068845-250MG
101937-912EA 729.22 USD
101937-912
2-Amino-4-methylthiophene ≥95%
2-Amino-4-methylthiophene

Hazard: Irritant

Formula: C₅H₇NS
CAS Number: 14770-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR