5-Amino-piperidin-2-one hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Aminopiperidin-2-one hydrochloride

068838-500MG
101937-888EA 600 USD
101937-888
5-Amino-piperidin-2-one hydrochloride ≥95%
5-Amino-piperidin-2-one hydrochloride

Hazard: Irritant

Formula: C₅H₁₁ClN₂O
MW: 150.61 g/mol
MDL Number: MFCD09743647
CAS Number: 1235440-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR