(6-Methoxy-3-pyridyl)methanol ≥95%

Supplier: Matrix Scientific

Synonyms: (6-Methoxypyridin-3-yl)methanol5-(Hydroxymethyl)-2-methoxypyridine

068836-500MG
101937-880EA 853.85 USD
101937-880
(6-Methoxy-3-pyridyl)methanol ≥95%
(6-Methoxy-3-pyridyl)methanol

Hazard: Irritant

Formula: C₇H₉NO₂
MW: 139.16 g/mol
MDL Number: MFCD06202826
CAS Number: 58584-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR