(6-Methyl-3-pyridyl)methanamine ≥95%

Supplier: Matrix Scientific

Synonyms: (6-Methylpyridin-3-yl)methanamine

068826-1G
101937-836EA 989.23 USD
101937-836
(6-Methyl-3-pyridyl)methanamine ≥95%
(6-Methyl-3-pyridyl)methanamine

Hazard: Irritant

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
MDL Number: MFCD06212657
CAS Number: 56622-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR