N-(6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-2-yl)methanesulfonamide ≥95%

Supplier: Matrix Scientific

068823-500MG
101937-824EA 1073.85 USD
101937-824
N-(6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-2-yl)methanesulfonamide ≥95%
N-(6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-2-yl)methanesulfonamide

Hazard: Irritant

Formula: C₁₇H₂₂BNO₄S
MDL Number: MFCD13191641
CAS Number: 1132940-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR