tert-Butyl-4-butylpiperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

068819-500MG
101937-816EA 524.62 USD
101937-816
tert-Butyl-4-butylpiperidine-1-carboxylate ≥95%
tert-Butyl-4-butylpiperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₂₇NO₂
MW: 241.38 g/mol
MDL Number: MFCD13191673
CAS Number: 1160592-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR