6-Bromo-2-naphthylamine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-6-bromonaphthalene6-Bromonaphthalen-2-amine

068816-5G
101937-798EA 80.5 USD
101937-798
6-Bromo-2-naphthylamine ≥95%
6-Bromo-2-naphthylamine

Hazard: Irritant

Formula: C₁₀H₈BrN
MW: 222.0812 g/mol
MDL Number: MFCD01026466
CAS Number: 7499-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR