3-(2-Methyl-1H-pyrrolo[2,3-b]pyridin-3-yl)propanoic acid ≥95%
Supplier: Matrix Scientific
068815-500MG
101937-794EA
1347.69
USD
101937-794
3-(2-Methyl-1H-pyrrolo[2,3-b]pyridin-3-yl)propanoic acid ≥95%
3-(2-Methyl-1H-pyrrolo[2,3-b]pyridin-3-yl)propanoic acid
Hazard: Irritant
Formula:
C₁₁H₁₂N₂O₂ |
MDL Number:
MFCD11040737 CAS Number: 18502-18-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...