tert-Butyl-5-oxo-4,9-diazaspiro[5.5]undecane-4-carboxylate ≥95%

Supplier: Matrix Scientific

068810-500MG
101937-782EA 1073.85 USD
101937-782
tert-Butyl-5-oxo-4,9-diazaspiro[5.5]undecane-4-carboxylate ≥95%
tert-Butyl-5-oxo-4,9-diazaspiro[5.5]undecane-4-carboxylate

Hazard: Irritant

Formula: C₁₄H₂₄N₂O₃
MW: 268.36 g/mol
MDL Number: MFCD16040216
CAS Number: 1221449-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR