2-Chloro-3-cyanopyrazine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-chloropyrazine-2-carbonitrile

068807-25G
101937-778EA 57.54 USD
101937-778
2-Chloro-3-cyanopyrazine ≥95%
2-Chloro-3-cyanopyrazine

Purity: 95%
CAS: 55557-52-3
MDL Number: MFCD00219653
Molecular Formula: C5H2ClN3
Molecular Weight: 139,54
Hazard: Irritant

Formula: C₅H₂ClN₃
MDL Number: MFCD00219653
CAS Number: 55557-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR