6-Bromo-1,4-dichlorophthalazine ≥95%
Supplier: Matrix Scientific
068805-500MG
101937-768EA
1073.85
USD
101937-768
6-Bromo-1,4-dichlorophthalazine ≥95%
6-Bromo-1,4-dichlorophthalazine
Hazard: Irritant
Formula:
C₈H₃BrCl₂N₂ |
MDL Number:
MFCD16037285 CAS Number: 240400-95-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...