4-Ethynyltetrahydro-2H-pyran ≥95%

Supplier: Matrix Scientific

068804-1G
101937-764EA 1130.77 USD
101937-764
4-Ethynyltetrahydro-2H-pyran ≥95%
4-Ethynyltetrahydro-2H-pyran

Hazard: Irritant

Formula: C₇H₁₀O
MDL Number: MFCD17016081
CAS Number: 1202245-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR