tert-Butyl-3-bromopyrazolo[3,4-b]pyridine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3-bromo-1H-pyrazolo[3,4-b]pyridine-1-carboxylate

068797-250MG
101937-730EA 524.62 USD
101937-730
tert-Butyl-3-bromopyrazolo[3,4-b]pyridine-1-carboxylate ≥95%
tert-Butyl-3-bromopyrazolo[3,4-b]pyridine-1-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₂BrN₃O₂
MW: 298.15 g/mol
MDL Number: MFCD17016052
CAS Number: 1234616-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR