Ethyl-5-chloro-1H-pyrrolo[2,3-b]pyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

068796-250MG
101937-726EA 815.38 USD
101937-726
Ethyl-5-chloro-1H-pyrrolo[2,3-b]pyridine-2-carboxylate ≥95%
Ethyl-5-chloro-1H-pyrrolo[2,3-b]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₉ClN₂O₂
MDL Number: MFCD11559021
CAS Number: 1083181-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR