6-Bromoimidazo[1,2-b]pyridazine ≥95%

Supplier: Matrix Scientific

068793-500MG
101937-712EA 933.85 USD
101937-712
6-Bromoimidazo[1,2-b]pyridazine ≥95%
6-Bromoimidazo[1,2-b]pyridazine

Hazard: Irritant

Formula: C₆H₄BrN₃
MDL Number: MFCD12198219
CAS Number: 1159977-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR