3-(4-Bromophenyl)isoxazole ≥95%

Supplier: Matrix Scientific

068788-5G
101937-688EA 880 USD
101937-688
3-(4-Bromophenyl)isoxazole ≥95%
3-(4-Bromophenyl)isoxazole

Hazard: Irritant

Formula: C₉H₆BrNO
MDL Number: MFCD08446384
CAS Number: 13484-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR