4-(Hydroxymethyl)piperidin-4-ol hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
4-(Hydroxymethyl)-4-piperidinol hydrochloride
068787-500MG
101937-684EA
576.92
USD
101937-684
4-(Hydroxymethyl)piperidin-4-ol hydrochloride ≥95%
4-(Hydroxymethyl)piperidin-4-ol hydrochloride
Hazard: Irritant
Formula:
C₆H₁₄ClNO₂ MW: 167.630 g/mol |
MDL Number:
MFCD11845427 CAS Number: 89584-31-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...