tert-Butyl (3R,4R)-3-Fluoro-4-hydroxypyrrolidine-1-carboxylate ≥96%

Supplier: Matrix Scientific

Synonyms: tert-butyl trans-3-fluoro-4-hydroxypyrrolidine-1-carboxylate

068786-500MG 068786-100MG
101937-676EA 349.23 USD
101937-676 101937-680
tert-Butyl (3R,4R)-3-Fluoro-4-hydroxypyrrolidine-1-carboxylate ≥96%
tert-Butyl (3R,4R)-3-Fluoro-4-hydroxypyrrolidine-1-carboxylate

Hazard: Irritant

Formula: C₉H₁₆FNO₃
MW: 205.23 g/mol
MDL Number: MFCD19443950
CAS Number: 869481-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR