5-Bromo-3-fluoro-2-(hydroxymethyl)pyridine ≥95%
Supplier: Matrix Scientific
Synonyms:
(5-Bromo-3-fluoropyridin-2-yl)methanol
068785-250MG
101937-672EA
1250.77
USD
101937-672
5-Bromo-3-fluoro-2-(hydroxymethyl)pyridine ≥95%
5-Bromo-3-fluoro-2-(hydroxymethyl)pyridine
Hazard: Irritant
Formula:
C₆H₅BrFNO MW: 206.02 g/mol |
MDL Number:
MFCD16607004 CAS Number: 1206968-92-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...