tert-Butyl-N-[(1-methyl-2-oxo-3-pyridyl)methyl]carbamate ≥95%

Supplier: Matrix Scientific

068769-500MG
101937-598EA 1538.46 USD
101937-598
tert-Butyl-N-[(1-methyl-2-oxo-3-pyridyl)methyl]carbamate ≥95%
tert-Butyl-N-[(1-methyl-2-oxo-3-pyridyl)methyl]carbamate

Hazard: Irritant

Formula: C₁₂H₁₈N₂O₃
MW: 238.29 g/mol
MDL Number: MFCD17214344
CAS Number: 1234616-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR