3-Hydroxy-1-isopropylazetidine hydrochloride ≥95%

Supplier: Matrix Scientific

068768-500MG
101937-594EA 660 USD
101937-594
3-Hydroxy-1-isopropylazetidine hydrochloride ≥95%
3-Hydroxy-1-isopropylazetidine hydrochloride

Hazard: Irritant

Formula: C₆H₁₄ClNO
MDL Number: MFCD00154136
CAS Number: 54431-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR