Methyl-8-bromoimidazo[1,2-a]pyridine-6-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 8-bromoimidazo[1,2-a]pyridine-6-carboxylate

068763-500MG
101937-574EA 933.85 USD
101937-574
Methyl-8-bromoimidazo[1,2-a]pyridine-6-carboxylate ≥95%
Methyl 8-bromoimidazo[1,2-a]pyridine-6-carboxylate

Hazard: Irritant

Formula: C₉H₇BrN₂O₂
MW: 255.0681 g/mol
MDL Number: MFCD17016026
CAS Number: 1234616-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR