Benzyl(1R,5S)-6-(hydroxymethyl)-3-azabicyclo[3.1.0]hexane-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: rel-Benzyl (1R,5S,6r)-6-(hydroxymethyl)-3-azabicyclo[3.1.0]hexane-3-carboxylate

068753-500MG
101937-530EA 1209.23 USD
101937-530
Benzyl(1R,5S)-6-(hydroxymethyl)-3-azabicyclo[3.1.0]hexane-3-carboxylate ≥95%
Benzyl(1R,5S)-6-(hydroxymethyl)-3-azabicyclo[3.1.0]hexane-3-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₇NO₃
MW: 247.3 g/mol
MDL Number: MFCD17214343
CAS Number: 134575-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR