tert-Butyl-N-[[(1S,5R)-3-azabicyclo[3.1.0]hexan-6-yl]methyl]carbamate ≥95%

Supplier: Matrix Scientific

068751-1G
101937-526EA 1535.38 USD
101937-526
tert-Butyl-N-[[(1S,5R)-3-azabicyclo[3.1.0]hexan-6-yl]methyl]carbamate ≥95%
tert-Butyl-N-[[(1S,5R)-3-azabicyclo[3.1.0]hexan-6-yl]methyl]carbamate

Hazard: Irritant

Formula: C₁₁H₂₀N₂O₂
MW: 212.29 g/mol
MDL Number: MFCD13189633
CAS Number: 134575-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR