Methyl-2-chloroquinazoline-7-carboxylate ≥95%

Supplier: Matrix Scientific

068749-500MG
101937-512EA 1787.69 USD
101937-512
Methyl-2-chloroquinazoline-7-carboxylate ≥95%
Methyl-2-chloroquinazoline-7-carboxylate

Hazard: Irritant

Formula: C₁₀H₇ClN₂O₂
MDL Number: MFCD12031313
CAS Number: 953039-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR