1H-Indazole-7-carbonitrile ≥95%

Supplier: Matrix Scientific

068744-250MG
101937-486EA 524.62 USD
101937-486
1H-Indazole-7-carbonitrile ≥95%
1H-Indazole-7-carbonitrile

Hazard: Irritant

Formula: C₈H₅N₃
MDL Number: MFCD10696804
CAS Number: 256228-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR