1-Methyl-7-azaindole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

068740-500MG
101937-470EA 1813.85 USD
101937-470
1-Methyl-7-azaindole-4-carboxylic acid ≥95%
1-Methyl-7-azaindole-4-carboxylic acid

Hazard: Irritant

Formula: C₉H₈N₂O₂
MDL Number: MFCD13302392
CAS Number: 1147753-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR