tert-Butyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrrolo[3,2-c]pyridine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[3,2-c]pyridine-1-carboxylate

068739-500MG
101937-466EA 1373.85 USD
101937-466
tert-Butyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrrolo[3,2-c]pyridine-1-carboxylate ≥95%
tert-Butyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrrolo[3,2-c]pyridine-1-carboxylate

Hazard: Irritant

Formula: C₁₈H₂₅BN₂O₄
MW: 344.22 g/mol
MDL Number: MFCD08234841
CAS Number: 877060-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR