1-(2-Tetrahydropyranyl)-1H-pyrazole-4-boronic acid pinacol ester ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(Tetrahydro-2H-pyran-2-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

068737-5G
101937-456EA 83.95 USD
101937-456
1-(2-Tetrahydropyranyl)-1H-pyrazole-4-boronic acid pinacol ester ≥95%
1-(2-Tetrahydropyranyl)-1H-pyrazole-4-boronic acid pinacol ester

Hazard: Irritant

Formula: C₁₄H₂₃BN₂O₃
MW: 278.1550 g/mol
MDL Number: MFCD16556148
CAS Number: 1003846-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR