3,5-Dichloro-4-hydroxybenzonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: Chloroxynil

068735-1G
101937-446EA 106.95 USD
101937-446
3,5-Dichloro-4-hydroxybenzonitrile ≥95%
3,5-Dichloro-4-hydroxybenzonitrile

Hazard: Irritant

Formula: C₇H₃Cl₂NO
MDL Number: MFCD00002177
CAS Number: 1891-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR