6-Iodo-1H-indazole ≥95%
Supplier: Matrix Scientific
Synonyms:
6-Iodoindazole
068734-1G
101937-442EA
150.77
USD
101937-442
6-Iodo-1H-indazole ≥95%
6-Iodo-1H-indazole
Hazard: Irritant
Formula:
C₇H₅IN₂ MW: 244.0300 g/mol |
MDL Number:
MFCD04114695 CAS Number: 261953-36-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...