2-Methylindazole-7-boronic acid pinacol ester ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-indazole

068730-500MG
101937-424EA 2692.31 USD
101937-424
2-Methylindazole-7-boronic acid pinacol ester ≥95%
2-Methylindazole-7-boronic acid pinacol ester

Hazard: Irritant

Formula: C₁₄H₁₉BN₂O₂
MW: 258.13 g/mol
MDL Number: MFCD11109457
CAS Number: 845751-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR