3-[Benzyl-(2-ethoxycarbonylethyl)amino]butyric acid ethyl ester ≥95%

Supplier: Matrix Scientific

068727-1G
101937-412EA 698.46 USD
101937-412
3-[Benzyl-(2-ethoxycarbonylethyl)amino]butyric acid ethyl ester ≥95%
3-[Benzyl-(2-ethoxycarbonylethyl)amino]butyric acid ethyl ester

Hazard: Irritant

Formula: C₁₈H₂₇NO₄
MDL Number: MFCD00480689
CAS Number: 58859-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR