4-Bromo-1-benzenesulfonyl-7-azaindole ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Benzenesulfonyl-4-bromo-7-azaindole1-Benzenesulphonyl-4-bromo-7-azaindole

068725-5G
101937-404EA 1892.31 USD
101937-404
4-Bromo-1-benzenesulfonyl-7-azaindole ≥95%
4-Bromo-1-benzenesulfonyl-7-azaindole

Hazard: Irritant

Formula: C₁₃H₉BrN₂O₂S
MW: 337.2 g/mol
MDL Number: MFCD08443800
CAS Number: 889939-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR