1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid pinacol ester ≥95%
Supplier: Matrix Scientific
Synonyms:
1-(Tetrahydro-2H-pyran-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
, 1-(Tetrahydro-2H-pyran-2-yl)-1H-pyrazole-5-boronic acid Pinacol ester
, 1-(Tetrahydropyran-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
068724-25G
101937-400EA
127.69
USD
101937-400
1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid pinacol ester ≥95%
1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid pinacol ester
Hazard: Irritant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...