tert-Butyl-4-ethynylpiperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-ethynylpiperidine-1-carboxylate

068721-1G
101937-384EA 60 USD
101937-384
tert-Butyl-4-ethynylpiperidine-1-carboxylate ≥95%
tert-Butyl-4-ethynylpiperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₉NO₂
MW: 209.2900 g/mol
MDL Number: MFCD09038026
CAS Number: 287192-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR