3-Amino-5-fluoro-1H-pyrazolo[3,4-b]pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Fluoro-1H-pyrazolo[3,4-b]pyridin-3-amine

068719-500MG
101937-374EA 1209.23 USD
101937-374
3-Amino-5-fluoro-1H-pyrazolo[3,4-b]pyridine ≥95%
3-Amino-5-fluoro-1H-pyrazolo[3,4-b]pyridine

Hazard: Irritant

Formula: C₆H₅FN₄
MW: 152.13 g/mol
MDL Number: MFCD15526909
CAS Number: 1034667-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR