[1,2,4]Triazolo[4,3-a]pyridine-7-carboxylic acid ≥95%

Supplier: Matrix Scientific

068715-250MG
101937-354EA 640 USD
101937-354
[1,2,4]Triazolo[4,3-a]pyridine-7-carboxylic acid ≥95%
[1,2,4]Triazolo[4,3-a]pyridine-7-carboxylic acid

Hazard: Irritant

Formula: C₇H₅N₃O₂
MDL Number: MFCD17015978
CAS Number: 1234616-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR