5-Bromo[1,2,4]triazolo[4,3-a]pyridine ≥95%

Supplier: Matrix Scientific

068714-1G
101937-350EA 1192.31 USD
101937-350
5-Bromo[1,2,4]triazolo[4,3-a]pyridine ≥95%
5-Bromo[1,2,4]triazolo[4,3-a]pyridine

Hazard: Irritant

Formula: C₆H₄BrN₃
MW: 198.03 g/mol
MDL Number: MFCD11043995
CAS Number: 1172085-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR