Methyl-3-hydroxypyridazine-6-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 6-oxo-1,6-dihydropyridazine-3-carboxylateMethyl 1,6-dihydro-6-oxopyridazine-3-carboxylate

068712-5G
101937-346EA 346.15 USD
101937-346
Methyl-3-hydroxypyridazine-6-carboxylate ≥95%
Methyl-3-hydroxypyridazine-6-carboxylate

Hazard: Irritant

Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
MDL Number: MFCD17015975
CAS Number: 63001-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR