Thieno[3,2-b]pyridine-2-carboxylic acid ≥95%
Supplier: Matrix Scientific
068710-500MG
101937-338EA
1209.23
USD
101937-338
Thieno[3,2-b]pyridine-2-carboxylic acid ≥95%
Thieno[3,2-b]pyridine-2-carboxylic acid
Hazard: Irritant
Formula:
C₈H₅NO₂S |
MDL Number:
MFCD12198115 CAS Number: 347187-30-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...