4-Indoleboronic acid pinacol ester ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

068707-5G
101937-324EA 138 USD
101937-324
4-Indoleboronic acid pinacol ester ≥95%
4-Indoleboronic acid pinacol ester

Hazard: Irritant

Formula: C₁₄H₁₈BNO₂
MW: 243.11 g/mol
MDL Number: MFCD08689896
CAS Number: 388116-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR