1H-Indazole-7-boronic acid pinacol ester ≥95%

Supplier: Matrix Scientific

Synonyms: 7-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

068702-5G
101937-302EA 686.55 USD
101937-302
1H-Indazole-7-boronic acid pinacol ester ≥95%
1H-Indazole-7-boronic acid pinacol ester

Hazard: Irritant

Formula: C₁₃H₁₇BN₂O₂
MW: 244.0973 g/mol
MDL Number: MFCD08059264
CAS Number: 915411-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR