3-Bromo-1H-indazole ≥95%

Supplier: Matrix Scientific

068701-5G
101937-298EA 85.1 USD
101937-298
3-Bromo-1H-indazole ≥95%
3-Bromo-1H-indazole

Hazard: Toxic

Formula: C₇H₅BrN₂
MW: 197.04 g/mol
Melting Pt: 148…149 °C
MDL Number: MFCD00159926
CAS Number: 40598-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR